Cal-520® alkyne
Lieferant: AAT BIOQUEST
Cal-520® alkyne is an excellent building block that can be readily used to prepare a calcium-sensitive bioconjugate for monitoring calcium change spatially for a specific target.
- Stable for 12 months
Calcium measurement is critical for numerous biological investigations.
Bitte beachten For Research Use Only