Tide Quencher™ 4 CPG 500 Å, Tide Quencher™ fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
TQ4 CPG
2062
AATB2062EA
273
EUR
AATB2062
Tide Quencher™ 4 CPG 500 Å, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 4 CPG
TQ4 is designed to be a superior quencher to ROX, TF4 and Texas Red®.
- Stronger absorption
- Higher quenching efficiency
- Versatile reactive forms
- Excellent oligonucleotide and peptide labeling
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...