Fura-8FF™ AM
Lieferant: AAT BIOQUEST
|
Achtung
|
20620
AATB20620EA
307
EUR
AATB20620
Fura-8FF™ AM
Fura-8FF™ AM
The cell-permeant Fura-8FF AM is an analog of Fura-8 AM with much lower calcium binding affinity, Kd ~10 µm.
- Calcium-dependent fluorescence
- Steady signal
- Cell permeant
Bitte beachten For research use only.
Molecular Weight:
973,86 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...