Fura-8FF™ pentapotassium salt

Lieferant: AAT BIOQUEST
Achtung

20621
AATB20621EA 307 EUR
AATB20621
Fura-8FF™ pentapotassium salt
Fura-8FF™ pentapotassium salt

The cell-impermeant Fura-8FF is an analog of Fura-8 with much lower calcium binding affinity, Kd ~10 µM.


  • Improved sensitivity
  • Low binding affinity
  • Long-wavelength emission
  • Cell impermeant


Bitte beachten For research use only.

Molecular Weight: 837,97 g/mol
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR