Tide Quencher™ 4 CPG 1000 Å, Tide Quencher™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TQ4 CPG

2063
AATB2063EA 273 EUR
AATB2063
Tide Quencher™ 4 CPG 1000 Å, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 4 CPG

TQ4 is designed to be a superior quencher to ROX, TF4 and Texas Red®.


  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling

ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR