Tide Quencher™ 4WS maleimide, Tide Quencher™ fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
TQ4WS maleimide
2064
AATB2064EA
460
EUR
AATB2064
Tide Quencher™ 4WS maleimide, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 4WS maleimide
TQ4WS is designed to be a superior quencher to ROX, TF4 and Texas Red®.
- Stronger absorption
- Higher quenching efficiency
- Versatile reactive forms
- Excellent oligonucleotide and peptide labeling
Molecular Weight:
1106,2 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...