Calbryte™-520L potassium salt

Lieferant: AAT BIOQUEST
Achtung

20642
AATB20642EA 460 EUR
AATB20642
Calbryte™-520L potassium salt
Calbryte™-520L potassium salt

The intracellular calcium flux assay is a widely used method for monitoring signal transduction pathways and high throughput screening of G protein-coupled receptors (GPCRs) and calcium channel targets.


  • Improved background/signal ratio
  • Low binding affinity
  • Cell impermeant


Bitte beachten For research use only.

Molecular Weight: 794,88 g/mol
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR