Calbryte™ 520 potassium salt
Lieferant: AAT BIOQUEST
20657
AATB20657EA
991
EUR
AATB20657
Calbryte™ 520 potassium salt
Calbryte™ 520 potassium salt
Calbryte™ 520 is a new generation of fluorescent indicator for calcium measurements; its greatly improved signal-to-noise ratio and better intracellular retention properties make Calbryte™ 520 the most robust calcium indicator.
- Stable for 12 months
Calcium measurement is critical for numerous biological investigations.
Bitte beachten For Research Use Only