Tide Quencher™ 4WS succinimidyl ester, Tide Quencher™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TQ4WS SE

2067
AATB2067EA 307 EUR
AATB2067
Tide Quencher™ 4WS succinimidyl ester, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 4WS succinimidyl ester

TQ4WS is designed to be a superior quencher to ROX, TF4 and Texas Red®.


  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling

Molecular Weight: 997,23 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2bda927fcb6d01adf52f36d0c6a21f5d4e44422c&vid=22a25dbf46664609b0332e2bf9f3f9b68b1fc59d&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR