Tide Quencher™ 4WS azide, Tide Quencher™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TQ4WS azide

2068
AATB2068EA 460 EUR
AATB2068
Tide Quencher™ 4WS azide, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 4WS azide

TQ4WS is designed to be a superior quencher to ROX, TF4 and Texas Red®.


  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling

Molecular Weight: 968,24 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85b22382e763079e41fcae536e130e14146f2da3&vid=41972cddd3104d5cd0f9162729a4bc040e135ac6&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR