4WS-DBCO [TQ4WS-DBCO], Tide Quencher™
Lieferant: AAT BIOQUEST
2070
AATB2070EA
429
EUR
AATB2070
4WS-DBCO [TQ4WS-DBCO], Tide Quencher™
4WS-DBCO [TQ4WS-DBCO]
TQ4WS is designed to be a superior quencher to ROX, TF4, iFluor® 594, Alexa Fluor® 594 and Texas Red®.
- Stable for 12 months
- Optimized for FRET applications
Quality tested in-house
Bitte beachten For Research Use Only