Calbryte™ 630 potassium salt
Lieferant: AAT Bioquest
Achtung
20727
AATB20727EA 460 EUR
AATB20727
Calbryte™ 630 potassium salt
Calbryte™ 630 potassium salt
Calcium measurement is critical for numerous biological investigations.
  • Improved background/signal ratio
  • Excellent intracellular retention
  • Long-wavelength emission
  • Cell impermeant
Bitte beachten For research use only.
ADR: 5,II