5.1WS acid [TQ5.1WS acid], Tide Quencher™
Lieferant: AAT BIOQUEST
2073
AATB2073EA
410
EUR
AATB2073
5.1WS acid [TQ5.1WS acid], Tide Quencher™
5.1WS acid [TQ5.1WS acid]
Tide Quencher™ 5.1WS (TQ5.1WS) is a non-fluorescent molecule designed to efficiently quench the fluorescence of common NIR fluorophores such as Cy5, Cy5.5, Alexa Fluor® 647, Alexa Fluor® 647, iFluor® 647 and iFluor® 680.
- Stable for 12 months
- Optimized for FRET applications
Quality tested in-house
Bitte beachten For Research Use Only