Tide Quencher™ 5WS acid, Tide Quencher™ fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
TQ5WS acid
2075
AATB2075EA
307
EUR
AATB2075
Tide Quencher™ 5WS acid, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 5WS acid
TQ5WS is designed to be a superior quencher to Cy5, TF5 and Cy5.5.
- Stronger absorption
- Higher quenching efficiency
- Versatile reactive forms
- Excellent oligonucleotide and peptide labeling
Molecular Weight:
890 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...