Tide Quencher™ 5WS amine, Tide Quencher™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TQ5WS amine

2076
AATB2076EA 307 EUR
AATB2076
Tide Quencher™ 5WS amine, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 5WS amine

TQ5WS is designed to be a superior quencher to Cy5, TF5 and Cy5.5.


  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling

Molecular Weight: 933,09 g/mol
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR