Tide Quencher™ 5 CPG 1000 Å, Tide Quencher™ fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
TQ5 CPG
2078
AATB2078EA
273
EUR
AATB2078
Tide Quencher™ 5 CPG 1000 Å, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 5 CPG
TQ5 is designed to be a superior quencher to Cy5, TF5 and Cy5.5.
- Stronger absorption
- Higher quenching efficiency
- Versatile reactive forms
- Excellent oligonucleotide and peptide labeling
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...