Tide Quencher™ 5WS maleimide, Tide Quencher™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TQ5WS maleimide

2079
AATB2079EA 460 EUR
AATB2079
Tide Quencher™ 5WS maleimide, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 5WS maleimide

TQ5WS is designed to be a superior quencher to Cy5, TF5 and Cy5.5.


  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling

Molecular Weight: 1083,2 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=453093476572971399063fde4115b093786dae97&vid=5521ef6aafd1934e07cbfff106eb3bd1bd209b32&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR