Tide Quencher™ 5WS succinimidyl ester, Tide Quencher™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TQ5WS SE

2081
AATB2081EA 307 EUR
AATB2081
Tide Quencher™ 5WS succinimidyl ester, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 5WS succinimidyl ester

TQ5WS is designed to be a superior quencher to Cy5, TF5 and Cy5.5.


  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling

Molecular Weight: 974,24 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf95d9f6911e0aa5934d31fc80decdb1c386a924&vid=061ff59ff2aaad4c56d27888321671db8c2c378e&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR