Tide Quencher™ 5WS azide, Tide Quencher™ fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
TQ5WS azide
2082
AATB2082EA
460
EUR
AATB2082
Tide Quencher™ 5WS azide, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 5WS azide
TQ5WS is designed to be a superior quencher to Cy5, TF5 and Cy5.5.
- Stronger absorption
- Higher quenching efficiency
- Versatile reactive forms
- Excellent oligonucleotide and peptide labeling
Molecular Weight:
958,02 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...