5.1 CPG [TQ5.1 CPG], Tide Quencher™ (500 A)
Lieferant: AAT BIOQUEST
2084
AATB2084EA 275 EUR
AATB2084
5.1 CPG [TQ5.1 CPG], Tide Quencher™ (500 A)
5.1 CPG [TQ5.1 CPG]
Tide Quencher™ 5.1 (TQ5.1) is a non-fluorescent molecule designed to efficiently quench the fluorescence of common NIR fluorophores such as Cy5, Cy5.5, Alexa Fluor® 647, Alexa Fluor® 647, iFluor® 647 and iFluor® 680.
  • Stable for 12 months
  • Optimized for FRET applications
Quality tested in-house
Bitte beachten For Research Use Only