5.1 CPG [TQ5.1 CPG], Tide Quencher™ 1000 Å
Lieferant: AAT BIOQUEST
2085
AATB2085EA
275
EUR
AATB2085
5.1 CPG [TQ5.1 CPG], Tide Quencher™ 1000 Å
5.1 CPG [TQ5.1 CPG]
Tide Quencher™ 5.1 (TQ5.1) is a non-fluorescent molecule designed to efficiently quench the fluorescence of common NIR fluorophores such as Cy5, Cy5.5, Alexa Fluor® 647, Alexa Fluor® 647, iFluor® 647 and iFluor® 680.
- Stable for 12 months
- Optimized for FRET applications
Quality tested in-house
Bitte beachten For Research Use Only