5.1WS azide [TQ5.1WS azide], Tide Quencher™
Lieferant: AAT BIOQUEST
2088
AATB2088EA 309 EUR
AATB2088
5.1WS azide [TQ5.1WS azide], Tide Quencher™
5.1WS azide [TQ5.1WS azide]
Tide Quencher™ 5.1WS (TQ5.1WS) is a non-fluorescent molecule designed to efficiently quench the fluorescence of common NIR fluorophores such as Cy5, Cy5.5, Alexa Fluor® 647, Alexa Fluor® 647, iFluor® 647 and iFluor® 680.
  • Stable for 12 months
  • Optimized for FRET applications
Quality tested in-house
Bitte beachten For Research Use Only