Tide Quencher™ 6WS amine, Tide Quencher™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TQ6WS amine

2091
AATB2091EA 460 EUR
AATB2091
Tide Quencher™ 6WS amine, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 6WS amine

TQ6WS is designed to be a superior quencher to Cy5.5, TF6 and Alexa Fluor 680.


  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling

Molecular Weight: 981,11 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b373a811385ca336cbfd385e02b037afd2552e8&vid=bf0d9516c7429d8f8a03d9f5e32f540012722cbc&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR