Tide Quencher™ 6WS maleimide, Tide Quencher™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TQ6WS maleimide

2094
AATB2094EA 460 EUR
AATB2094
Tide Quencher™ 6WS maleimide, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 6WS maleimide

TQ6WS is designed to be a superior quencher to Cy5.5, TF6 and Alexa Fluor 680.


  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling

Molecular Weight: 1132,24 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c0dac0a0a7c277d862afbc2d0a609e5955032298&vid=fca9f2eafdecb2aed1de24c44512ec6672c7f270&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR