Tide Quencher™ 6WS alkyne, Tide Quencher™ fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
TQ6WS alkyne
2098
AATB2098EA
460
EUR
AATB2098
Tide Quencher™ 6WS alkyne, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 6WS alkyne
TQ6WS is designed to be a superior quencher to Cy5.5, TF6 and Alexa Fluor 680.
- Stronger absorption
- Higher quenching efficiency
- Versatile reactive forms
- Excellent oligonucleotide and peptide labeling
Molecular Weight:
976,1 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...