BAPTA-AM ≥95%

Lieferant: AAT Bioquest
Achtung

21001
AATB21001EA 120 EUR
AATB21001
BAPTA-AM ≥95%
BAPTA-AM

BAPTA AM is cell-permeable version of BAPTA (1,2-bis(o-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid), a cell-permeable calcium chelator.

Formel: C₃₄H₄₀N₂O₁₈
MDL: MFCD00036696
CAS-Nummer: 126150-97-8
ADR: 5,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR