BAPTA tetrapotassium salt

Lieferant: AAT BIOQUEST

Synonyme: BAPTA 4K, 1,2-Bis(2-aminophenoxy)ethane-N,N,N′,N′-tetraacetic acid tetrapotassium salt, Potassium 2,2',2'',2'''-(((ethane-1,2-diylbis(oxy))bis(2,1-phenylene))bis(azanetriyl))tetraacetate

21003
AATB21003EA 120 EUR
AATB21003
BAPTA tetrapotassium salt
BAPTA tetrapotassium salt

BAPTA (1,2-bis(o-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid) is a water-soluble and cell-impermeable calcium chelator.

Formel: C₂₂H₂₀N₂O₁₀K₄
Molecular Weight: 628,79 g/mol
MDL: MFCD00070080
CAS-Nummer: 73630-08-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR