BAPTA tetrasodium salt

Lieferant: AAT BIOQUEST
Achtung

Synonyme: 1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid tetrasodium Salt, Tetrasodium-1,2-bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetate, BAPTA-Na4

21004
AATB21004EA 120 EUR
AATB21004
BAPTA tetrasodium salt
BAPTA tetrasodium salt

BAPTA (1,2-bis(o-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid) is a water-soluble and cell-impermeable calcium chelator.

Formel: C₂₂H₂₄N₂O₁₀
Molecular Weight: 564.37 g/mol
MDL: MFCD00067691
CAS-Nummer: 126824-24-6
ADR: 5 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR