EGTA-Na4 (Ethylenglycolbis(2-aminoethylether)-N,N,N',N'-tetraessigsäureTetranatriumsalz) ≥95%
Lieferant: AAT BIOQUEST
|
Achtung
|
21007
AATB21007EA
120
EUR
AATB21007
EGTA-Na4 (Ethylenglycolbis(2-aminoethylether)-N,N,N',N'-tetraessigsäureTetranatriumsalz) ≥95%
EGTA-Na4 (Ethylenglycolbis(2-aminoethylether)-N,N,N',N'-tetraessigsäureTetranatriumsalz)
EGTA (1,2-bis(o-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid) is a water-soluble and cell-impermeable calcium chelator.
Formel:
C₁₄H₂₀N₂Na₄O₁₀ |
MDL:
MFCD00274420 CAS-Nummer: 13368-13-3 ADR: 5,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...