EGTA-Na4 (Ethylenglycolbis(2-aminoethylether)-N,N,N',N'-tetraessigsäureTetranatriumsalz) 10 mM Kalziumchelatbildner

Lieferant: AAT BIOQUEST
Achtung

21008
AATB21008EA 120 EUR
AATB21008
EGTA-Na4 (Ethylenglycolbis(2-aminoethylether)-N,N,N',N'-tetraessigsäureTetranatriumsalz) 10 mM Kalziumchelatbildner
EGTA-Na4 (Ethylenglycolbis(2-aminoethylether)-N,N,N',N'-tetraessigsäureTetranatriumsalz)

EGTA (1,2-bis(o-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid) is a water-soluble and cell-impermeable calcium chelator.


  • High selectivity
  • Cell permeable

Formel: C₁₄H₂₀N₂Na₄O₁₀
MDL: MFCD00274420
CAS-Nummer: 13368-13-3
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=935bb9c106d54859a9e6a70b482995d6ba65062f&vid=315e4ae84ff7a8c8821eaa0651f186f114bd5095&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR