FLUO 3/AM ≥95%, Ultrareine Qualität
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
4-(6-Acetoxymethoxy-2,7-dichloro-3-oxo-9-xanthenyl)-4′-methyl-2,2′(ethylenedioxy)dianiline-N,N,N′,N′-tetraacetic acid tetrakis(acetoxymethyl) ester, 1-[Amino-5-(2,7-dichloro-6-acetomethoxy-3-oxo-3H-xanthen-9-yl)phenoxy]-2-(2'-amino-5'-methylphenoxy)ethane-N,N,N',N'-tetraacetic acid pentaacetoxy-methyl ester
21011
21013
AATB21011EA
273
EUR
AATB21011
AATB21013
FLUO 3/AM ≥95%, Ultrareine Qualität
FLUO 3/AM
Calcium measurement is critical for numerous biological investigations.
Formel:
C₅₁H₅₀Cl₂N₂O₂₃ Molecular Weight: 1129.85 g/mol |
CAS-Nummer:
121714-22-5 ADR: 5 II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...