FLUO-3 pentapotassium salt

Lieferant: AAT BIOQUEST
Achtung

21017
AATB21017EA 230 EUR
AATB21017
FLUO-3 pentapotassium salt
FLUO-3 pentapotassium salt

Calcium measurement is critical for numerous biological investigations.


  • Well-cited
  • Suitable for all major fluorescence platforms
  • Cell impermeant


Bitte beachten For research use only.

Formel: C₃₆H₂₅Cl₂K₅N₂O₁₃
Molecular Weight: 960 g/mol
CAS-Nummer: 134907-84-9
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR