FURA-2 pentapotassium salt ≥95%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: 1-[2-(5-Carboxyoxazol-2-yl)-6-aminobenzofuran-5-oxy]-2-(2'-amino-5'-methyl-phenoxy)ethane-N,N,N',N'-tetraacetic acid . 5K, FURA-2

21025
AATB21025EA 152 EUR
AATB21025
FURA-2 pentapotassium salt ≥95%
FURA-2 pentapotassium salt

Among the ratiometric calcium indicators, Fura-2 and Indo-1 are most commonly used.

Formel: C₂₉H₂₂K₅N₃O₁₄
Molecular Weight: 832 g/mol
CAS-Nummer: 113694-64-7
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR