FURA-2 pentasodium salt
Lieferant: AAT BIOQUEST
|
Achtung
|
21026
AATB21026EA
152
EUR
AATB21026
FURA-2 pentasodium salt
FURA-2 pentasodium salt
Among the ratiometric calcium indicators, Fura-2 and Indo-1 are most commonly used.
- Calcium-dependent fluorescence
- Ratiometric detection
- Cell impermeant
Bitte beachten For research use only.
Formel:
C₂₉H₂₂N₃Na₅O₁₄ Molecular Weight: 751,45 g/mol |
MDL:
MFCD00083328 CAS-Nummer: 96314-98-6 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...