Indo-1 AM ≥95%

Lieferant: AAT Bioquest
Achtung

21030 21033
AATB21030EA 152 EUR
AATB21030 AATB21033
Indo-1 AM ≥95%
Indo-1 AM

Calcium measurements are critical for numerous biological investigations.

Formel: C₄₇H₅₁N₃O₂₂
MDL: MFCD00036955
CAS-Nummer: 112926-02-0
ADR: 5,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR