Indo-1 pentapotassium salt ≥95%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: Indo-1 5K⁺

21040
AATB21040EA 152 EUR
AATB21040
Indo-1 pentapotassium salt ≥95%
Indo-1 pentapotassium salt

Calcium measurements are critical for numerous biological investigations.

Formel: C₃₂H₂₆K₅N₃O₁₂
Molecular Weight: 840,05 g/mol
CAS-Nummer: 132319-56-3
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR