Indo-1 pentasodium salt ratiometrischer Calciumindikator

Lieferant: AAT BIOQUEST
Achtung

Synonyme: Indo-1 5Na⁺

21044
AATB21044EA 152 EUR
AATB21044
Indo-1 pentasodium salt ratiometrischer Calciumindikator
Indo-1 pentasodium salt

Calcium measurements are critical for numerous biological investigations.


  • Ratiometric calcium indicator
  • Optimized for flow cytometry

Formel: C₃₂H₂₆N₃Na₅O₁₂
Molecular Weight: 759,5 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=61b3f25f9bc8eed79526c9bfb1bec055e406c9f2&vid=5fb51c6751fa7fbbe2c30d02114710ca09f69fe7&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR