Tide Quencher™ 7WS acid, Tide Quencher™ fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
TQ7WS acid
2105
AATB2105EA
769
EUR
AATB2105
Tide Quencher™ 7WS acid, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 7WS acid
TQ7WS is designed to be a superior quencher to Cy7, TF7 and Alexa Fluor 750.
- Stronger absorption
- Higher quenching efficiency
- Versatile reactive forms
- Excellent oligonucleotide and peptide labeling
Molecular Weight:
916,03 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...