Quin 2-Am ≥95%

Lieferant: AAT BIOQUEST
Achtung

21050
AATB21050EA 273 EUR
AATB21050
Quin 2-Am ≥95%
Quin 2-Am

Quin-2 binds calcium tightly and resembles calcium chelator EGTA in ability to bind calcium much more tightly than magnesium.

Formel: C₃₈H₄₃N₃O₁₈
Molecular Weight: 829.765 g/mol
CAS-Nummer: 83104-85-2
ADR: 5 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR