Quin 2 potassium salt hydrate ≥95%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: 2-([2-Bis(carboxymethyl)amino-5-methylphenoxy]methyl)-6-methoxy-8-bis(carboxymethyl)aminoquinoline

21052
AATB21052EA 230 EUR
AATB21052
Quin 2 potassium salt hydrate ≥95%
Quin 2 potassium salt hydrate

Quin-2 binds calcium tightly and resembles calcium chelator EGTA in ability to bind calcium much more tightly than magnesium.

Formel: C₂₆H₂₇N₃O
MDL: MFCD00167201
CAS-Nummer: 149022-19-5
ADR: 5,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR