BTC, tetrapotassium salt ≥95%
Lieferant: AAT BIOQUEST
|
Achtung
|
21053
AATB21053EA
307
EUR
AATB21053
BTC, tetrapotassium salt ≥95%
BTC, tetrapotassium salt
This cell-impermeant coumarin Ca²⁺ indicator exhibits a shift in excitation maximum from about 480 to 400 nm upon binding Ca²⁺, enabling ratiometric calcium measurements.
Formel:
C₃₃H₂₅N₃O₁₂S·K₄ Molecular Weight: 844,02 g/mol |
CAS-Nummer:
216453-54-2 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...