BTC, AM ≥95%
Lieferant: AAT Bioquest
|
Achtung
|
21054
AATB21054EA
383
EUR
AATB21054
BTC, AM ≥95%
BTC, AM
This cell-permeant coumarin Ca2+ indicator BTC, AM exhibits a shift in excitation maximum from about 480 nm to 400 nm upon binding Ca2+, enabling ratiometric calcium measurements.
Formel:
C₄₅H₄₅N₃O₂₀S |
CAS-Nummer:
176767-94-5 ADR: 5,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...