Fura-8™ pentapotassium salt

Lieferant: AAT BIOQUEST
Achtung

21057
AATB21057EA 152 EUR
AATB21057
Fura-8™ pentapotassium salt
Fura-8™ pentapotassium salt

Although Fura-2 has become the preferred excitation-ratioable calcium indicator of choice, it has certain limitations (e.g., lower sensitivity than single wavelength calcium indicators such as Fluo-8® and Cal-520®).


  • Improved background/signal ratio
  • Excellent intracellular retention
  • Long-wavelength emission
  • Cell impermeant


Bitte beachten For research use only.

Molecular Weight: 816,01 g/mol
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR