Tide Quencher™ 7WS amine, Tide Quencher™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TQ7WS amine

2106
AATB2106EA 614 EUR
AATB2106
Tide Quencher™ 7WS amine, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 7WS amine

TQ7WS is designed to be a superior quencher to Cy7, TF7 and Alexa Fluor 750.


  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling

Molecular Weight: 1072,14 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=410854140e5e0ecc5dc0317f75e192f9fb723b6a&vid=a8fa1943f69efba389797450a9bf2acdc68f47f0&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR