Rhod-2 AM (Bromide) ≥90%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: Rhod-2 AM ester, 1-[2-Amino-5-(3-dimethylamino-6-dimethylammonio-9-xanthenyl)phenoxy]-2-(2-amino-5-methylphenoxy)]ethane-N,N,N',N'-tetraacetic acid, tetraacetoxy methyl ester . Br

21060
AATB21060EA 273 EUR
AATB21060
Rhod-2 AM (Bromide) ≥90%
Rhod-2 AM (Bromide)

Calcium measurement is critical for numerous biological investigations.

Formel: C₅₂H₅₉BrN₄O₁₉
Molecular Weight: 1124,0 g/mol
CAS-Nummer: 145037-81-6
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR