Rhod-2, tripotassium salt Kalziumindikator
Lieferant: AAT BIOQUEST
|
Achtung
|
21067
AATB21067EA
307
EUR
AATB21067
Rhod-2, tripotassium salt Kalziumindikator
Rhod-2, tripotassium salt
Calcium measurement is critical for numerous biological investigations.
- Intracellular probe
- Calcium-dependent fluorescence
Molecular Weight:
869,05 g/mol |
CAS-Nummer:
663625-18-1 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...