Rhod-5N tripotassium salt

Lieferant: AAT BIOQUEST
Achtung

21072
AATB21072EA 383 EUR
AATB21072
Rhod-5N tripotassium salt
Rhod-5N tripotassium salt

Calcium measurement is critical for numerous biological investigations.


  • Calcium-dependent fluorescence
  • Low binding affinity
  • Cell impermeant


Bitte beachten For research use only.

Molecular Weight: 900.02 g/mol
ADR: 5 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR