Tide Quencher™ 7WS maleimide, Tide Quencher™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TQ7WS maleimide

2109
AATB2109EA 614 EUR
AATB2109
Tide Quencher™ 7WS maleimide, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 7WS maleimide

TQ7WS is designed to be a superior quencher to Cy7, TF7 and Alexa Fluor 750.


  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling

Molecular Weight: 1109.24 g/mol
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75f5cfb9466ffbba96c6f2ef0d48f0058c5d0198&vid=4b6fedfa5b5f2cc2035a17deb7cadbd6fda4338f&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR