Fluo-8H™ sodium salt
Lieferant: AAT Bioquest
Achtung
21095
AATB21095EA 307 EUR
AATB21095
Fluo-8H™ sodium salt
Fluo-8H™ sodium salt
Calcium measurements are critical for numerous biological investigations.
  • Can be loaded at rt
  • Improved calcium response
  • Significantly increased signal
  • Cell impermeant
  • Suitable for all fluorescence platforms
Bitte beachten For research use only.
ADR: 5,II