Fluo-8H™ sodium salt
Lieferant: AAT Bioquest
|
Achtung
|
21095
AATB21095EA
307
EUR
AATB21095
Fluo-8H™ sodium salt
Fluo-8H™ sodium salt
Calcium measurements are critical for numerous biological investigations.
- Can be loaded at rt
- Improved calcium response
- Significantly increased signal
- Cell impermeant
- Suitable for all fluorescence platforms
Bitte beachten For research use only.
ADR:
5,II |