6-Carboxy-2',4,7',7-tetrachlorofluorescein succinimidyl ester (6-TET SE) ≥90%
Lieferant: AAT BIOQUEST
Synonyme: 6-TET, NHS ester
211
AATB211EA 230 EUR
AATB211
6-Carboxy-2',4,7',7-tetrachlorofluorescein succinimidyl ester (6-TET SE) ≥90%
6-Carboxy-2',4,7',7-tetrachlorofluorescein succinimidyl ester (6-TET SE)
6-TET, SE is a popular amino-reactive fluorescent probe that is widely used in nucleic acid sequencing and related research.
  • Useful for Genetic Research
  • Excellent Quality
Bitte beachten For research use only.